Anti-EIF4B Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB38359-100EA 657 EUR
ABCAAB38359-100
Anti-EIF4B Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: EIF4B (eukaryotic translation initiation factor 4B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR