Anti-ITGB3 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB38460-100EA 688 EUR
ABCAAB38460-100
Anti-ITGB3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ITGB3 (integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR