Anti-CAV1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB38468-100EA 688 EUR
ABCAAB38468-100
Anti-CAV1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CAV1 (caveolin 1, caveolae protein, 22kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR