Anti-AKT2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB38513-100EA 713 EUR
ABCAAB38513-100
Anti-AKT2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AKT2 (v-akt murine thymoma viral oncogene homolog 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR