Anti-NFKBIA Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB38515-100EA 697 EUR
ABCAAB38515-100
Anti-NFKBIA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NFKBIA (nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR