Anti-NR6A1 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB38816-200EA 853 EUR
ABCAAB38816-200
Anti-NR6A1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NR6A1 (nuclear receptor subfamily 6, group A, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat