Anti-TBX6 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB38883-100EA 914 EUR
ABCAAB38883-100
Anti-TBX6 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TBX6 (T-box 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR