Anti-MMP9 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB38898-100EA 738 EUR
ABCAAB38898-100
Anti-MMP9 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MMP9 (matrix metallopeptidase 9 (gelatinase B, 92kDa gelatinase, 92kDa type IV collagenase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR