Anti-Plasmodium aldolase Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Lieferant: Abcam
ABCAAB38905-250EA 877 EUR
ABCAAB38905-250
Anti-Plasmodium aldolase Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Type: Primary
Antigen: Plasmodium aldolase
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen