Anti-MMP1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB38929-100EA 938 EUR
ABCAAB38929-100
Anti-MMP1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MMP1 (matrix metallopeptidase 1 (interstitial collagenase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR