Anti-PKA R2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB38949-100EA 688 EUR
ABCAAB38949-100
Anti-PKA R2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PKA R2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR