Anti-MMP14 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB38971-100EA 719 EUR
ABCAAB38971-100
Anti-MMP14 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MMP14 (matrix metallopeptidase 14 (membrane-inserted))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: Hinge region
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR