Anti-TIMP4 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB38987-100EA 671 EUR
ABCAAB38987-100
Anti-TIMP4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TIMP4 (TIMP metallopeptidase inhibitor 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR