Anti-PLK1 Mouse Monoclonal Antibody [clone: 2A3]

Lieferant: Abcam
ABCAAB39068-100EA 703 EUR
ABCAAB39068-100
Anti-PLK1 Mouse Monoclonal Antibody [clone: 2A3]
Antikörper
Type: Primary
Antigen: PLK1 (polo-like kinase 1)
Clonality: monoclonal
Clone: 2A3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR