Anti-PRC1 Mouse Monoclonal Antibody [clone: 6G2]

Lieferant: Abcam
ABCAAB39069-100EA 882 EUR
ABCAAB39069-100
Anti-PRC1 Mouse Monoclonal Antibody [clone: 6G2]
Antikörper
Type: Primary
Antigen: PRC1 (protein regulator of cytokinesis 1)
Clonality: monoclonal
Clone: 6G2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR