Anti-ADAM12 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB39155-100EA 697 EUR
ABCAAB39155-100
Anti-ADAM12 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ADAM12 (ADAM metallopeptidase domain 12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Pig, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR