Anti-CAPN2 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB39165-100EA 930 EUR
ABCAAB39165-100
Anti-CAPN2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CAPN2 (calpain 2, (m/II) large subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat