Anti-ACE Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB39172-100EA 682 EUR
ABCAAB39172-100
Anti-ACE Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ACE (angiotensin I converting enzyme)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR