Anti-ADAM10 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB39177-100EA 688 EUR
ABCAAB39177-100
Anti-ADAM10 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ADAM10 (ADAM metallopeptidase domain 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: cytoplasmic domain
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR