Anti-TIMP3 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB39184-100EA 719 EUR
ABCAAB39184-100
Anti-TIMP3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TIMP3 (TIMP metallopeptidase inhibitor 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: Loop 1
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR