Anti-ADAMTS5 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB39202-100EA 703 EUR
ABCAAB39202-100
Anti-ADAMTS5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ADAMTS5 (ADAM metallopeptidase with thrombospondin type 1 motif, 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR