Anti-PCOLCE Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB39204-100EA 896 EUR
ABCAAB39204-100
Anti-PCOLCE Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PCOLCE (procollagen C-endopeptidase enhancer)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR