Anti-MGMT Mouse Monoclonal Antibody [clone: MT3.1]

Lieferant: Abcam
ABCAAB39253-500EA 779 EUR
ABCAAB39253-500
Anti-MGMT Mouse Monoclonal Antibody [clone: MT3.1]
Antikörper
Type: Primary
Antigen: MGMT (O-6-methylguanine-DNA methyltransferase)
Clonality: monoclonal
Clone: MT3.1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR