Anti-ALOX5 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB39347-500EA 688 EUR
ABCAAB39347-500
Anti-ALOX5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ALOX5 (arachidonate 5-lipoxygenase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR