Anti-PLXNA4 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB39350-100EA 944 EUR
ABCAAB39350-100
Anti-PLXNA4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PLXNA4 (plexin A4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat