Anti-ZBTB16 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB39354-100EA 682 EUR
ABCAAB39354-100
Anti-ZBTB16 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZBTB16 (zinc finger and BTB domain containing 16)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR