Anti-AVP Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB39363-50EA 682 EUR
ABCAAB39363-50
Anti-AVP Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AVP (arginine vasopressin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR