Anti-NSE Chicken Polyclonal Antibody

Lieferant: Abcam
ABCAAB39369-100EA 688 EUR
ABCAAB39369-100
Anti-NSE Chicken Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NSE
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR