Anti-NTN1 Chicken Polyclonal Antibody

Lieferant: Abcam
ABCAAB39370-100EA 688 EUR
ABCAAB39370-100
Anti-NTN1 Chicken Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NTN1 (netrin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR