Anti-PRPH Chicken polyclonal antibody

Lieferant: Abcam
ABCAAB39374-100EA 644 EUR
ABCAAB39374-100
Anti-PRPH Chicken polyclonal antibody
Type: Primary
Antigen: PRPH (peripherin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR