Anti-IGF1R Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB39398-100EA 719 EUR
ABCAAB39398-100
Anti-IGF1R Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IGF1R (insulin-like growth factor 1 receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR