Anti-MAP3K5 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB39402-100EA 882 EUR
ABCAAB39402-100
Anti-MAP3K5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MAP3K5 (mitogen-activated protein kinase kinase kinase 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse