Anti-INA Mouse Monoclonal Antibody [clone: 1D2]

Lieferant: Abcam
ABCAAB39432-100EA 666 EUR
ABCAAB39432-100
Anti-INA Mouse Monoclonal Antibody [clone: 1D2]
Antikörper
Type: Primary
Antigen: INA (internexin neuronal intermediate filament protein, alpha)
Clonality: monoclonal
Clone: 1D2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR