Anti-ALOX5AP Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB39535-100EA 690 EUR
ABCAAB39535-100
Anti-ALOX5AP Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ALOX5AP (arachidonate 5-lipoxygenase-activating protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR