Anti-TYK2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB39550-100EA 672 EUR
ABCAAB39550-100
Anti-TYK2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TYK2 (tyrosine kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR