Anti-C1S Mouse Monoclonal Antibody [clone: 9]

Lieferant: Abcam
ABCAAB39551-100EA 672 EUR
ABCAAB39551-100
Anti-C1S Mouse Monoclonal Antibody [clone: 9]
Antikörper
Type: Primary
Antigen: C1S (complement component 1, s subcomponent)
Clonality: monoclonal
Clone: 9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR