Anti-SF3B1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB39578-100EA 899 EUR
ABCAAB39578-100
Anti-SF3B1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SF3B1 (splicing factor 3b, subunit 1, 155kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR