Anti-IgM Goat Polyclonal Antibody (Gold, 6nm)

Lieferant: Abcam
ABCAAB39606-1250EA 494 EUR
ABCAAB39606-1250
Anti-IgM Goat Polyclonal Antibody (Gold, 6nm)
Antikörper
Type: Secondary
Antigen: IgM (Mouse) (mouse IgM)
Clonality: polyclonal
Clone:
Conjugation: 6 nm Gold
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR