Anti-IgG Rabbit Polyclonal Antibody (Gold, 6nm)

Lieferant: Abcam
ABCAAB39610-1250EA 517 EUR
ABCAAB39610-1250
Anti-IgG Rabbit Polyclonal Antibody (Gold, 6nm)
Antikörper
Type: Secondary
Antigen: IgG (Goat) (goat IgG)
Clonality: polyclonal
Clone:
Conjugation: 6 nm Gold
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR