Anti-CCL8 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB39625-100EA 690 EUR
ABCAAB39625-100
Anti-CCL8 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: CCL8 (chemokine (C-C motif) ligand 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR