Anti-CCL8 Rabbit Polyclonal Antibody (Biotin)

Lieferant: Abcam
ABCAAB39627-50EA 666 EUR
ABCAAB39627-50
Anti-CCL8 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: CCL8 (chemokine (C-C motif) ligand 8)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR