Anti-HTR3B Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB39629-200EA 914 EUR
ABCAAB39629-200
Anti-HTR3B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HTR3B (5-hydroxytryptamine (serotonin) receptor 3B, ionotropic)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR