Anti-JAK2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB39636-100EA 688 EUR
ABCAAB39636-100
Anti-JAK2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: JAK2 (Janus kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR