Anti-HAP1 Mouse monoclonal antibody unconjugated [clone: 1B6]

Lieferant: Abcam
ABCAAB39713-100EA 634 EUR
ABCAAB39713-100
Anti-HAP1 Mouse monoclonal antibody unconjugated [clone: 1B6]
Antikörper
Type: Primary
Antigen: HAP1 (huntingtin-associated protein 1)
Clonality: monoclonal
Clone: 1B6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR