Anti-Hepatitis B Virus X antigen Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB39716-50EA 672 EUR
ABCAAB39716-50
Anti-Hepatitis B Virus X antigen Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Hepatitis B Virus X antigen
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR