Anti-PLXNB1 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB39717-100EA 896 EUR
ABCAAB39717-100
Anti-PLXNB1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PLXNB1 (plexin B1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat