Anti-G-protein coupled receptor 30 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB39742-100EA 909 EUR
ABCAAB39742-100
Anti-G-protein coupled receptor 30 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: G-protein coupled receptor 30
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR