Anti-PCSK5 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB39873-100EA 688 EUR
ABCAAB39873-100
Anti-PCSK5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PCSK5 (proprotein convertase subtilisin/kexin type 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR