Anti-GRPR Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB39883-100EA 908 EUR
ABCAAB39883-100
Anti-GRPR Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GRPR (gastrin-releasing peptide receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR