Anti-CALR Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB39897-200EA 749 EUR
ABCAAB39897-200
Anti-CALR Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CALR (calreticulin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR